missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hexafluorophosphate, 99%

Catalog No. AAA1746722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1746722 100 g
AAA1746714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1746722 Supplier Thermo Scientific Chemicals Supplier No. A1746722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 558-32-7 | C4H12F6NP | 219.111 g/mol

It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

Chemical Identifiers

CAS 558-32-7
Molecular Formula C4H12F6NP
Molecular Weight (g/mol) 219.111
MDL Number MFCD00012012
InChI Key ZFKULDQWLXAKIM-UHFFFAOYSA-N
Synonym tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
PubChem CID 11209
IUPAC Name tetramethylazanium;hexafluorophosphate
SMILES C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point >300°C
Odor Odorless
Linear Formula (CH3)4NPF6
Quantity 100 g
UN Number UN2923
Beilstein 3730394
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 219.1
Percent Purity ≥99%
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less
Hazard Category H301-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 209-194-3
RTECSNumber BS8000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only