missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Dichloroiodate 95.0+%, TCI America™

Catalog No. t27175g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27175G Supplier TCI America Supplier No. T27175G

Chemical Identifiers

CAS 1838-41-1
Molecular Formula C4H12Cl2IN
Molecular Weight (g/mol) 271.95
MDL Number MFCD00800939
InChI Key FUNMSHCNGVSXHD-UHFFFAOYSA-N
PubChem CID 11737348
IUPAC Name dichloroiodanuide; tetramethylazanium
SMILES Cl[I-]Cl.C[N+](C)(C)C

Specifications

Melting Point 216°C
Color Yellow
Quantity 5 g
Formula Weight 271.95
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Dichloroiodate
EINECSNumber (2)-0186
TSCA No