missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Bromide 97.0+%, TCI America™

Catalog No. T013525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013525G 25 g
T0135500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T013525G Supplier TCI America Supplier No. T013525G

Chemical Identifiers

CAS 64-20-0
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD00011626
InChI Key DDFYFBUWEBINLX-UHFFFAOYSA-M
Synonym tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm
PubChem CID 66137
ChEBI CHEBI:55317
IUPAC Name tetramethylazanium bromide
SMILES [Br-].C[N+](C)(C)C

Specifications

Color White
pH 4.0 to 7.0 (50g/L H2O soln.)
Quantity 25 g
UN Number 2811
Formula Weight 154.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Bromide
EINECSNumber (2)-0186
RTECSNumber BS7600000
TSCA Yes