missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium bromide, 98%

Catalog No. AAA1336822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1336822 100 g
AAA1336836 500 g
AAA133680E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1336822 Supplier Thermo Scientific Chemicals Supplier No. A1336822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 64-20-0 | C4H12BrN | 154.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetramethylammonium bromide acts as an ion pair reagent. It is used as a surface-active agent and an active ingredient for conditioners. Further, it is used as a softner for textiles and paper products. It is also employed as a disinfection agent, sanitizer, antimicrobial and antistatic agent. In addition to this, it plays in important role as a phase transfer catalyst in synthetic chemistry.

Solubility
Soluble in water, alcohol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with oxidizing agents and strong bases.

Chemical Identifiers

CAS 64-20-0
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD00011626
InChI Key DDFYFBUWEBINLX-UHFFFAOYSA-M
Synonym tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm
PubChem CID 66137
ChEBI CHEBI:55317
IUPAC Name tetramethylazanium;bromide
SMILES [Br-].C[N+](C)(C)C

Specifications

Melting Point >300°C
Density 1.56
Odor Odorless
Linear Formula (CH3)4NBr
Quantity 100 g
UN Number UN2811
Beilstein 3620955
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,chloroform and acetone.
Formula Weight 154.06
Percent Purity 98%
Chemical Name or Material Tetramethylammonium bromide
Show More Show Less
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-581-2
RTECSNumber BS7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only