missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Borohydride 90.0+%, TCI America™
$65.24 - $184.54
Chemical Identifiers
| CAS | 16883-45-7 |
|---|---|
| Molecular Formula | C4H12BN |
| Molecular Weight (g/mol) | 84.96 |
| MDL Number | MFCD00011778 |
| InChI Key | TVQBZLPZXCVNEO-UHFFFAOYSA-N |
| Synonym | tetramethylammonium borohydride, borohydride, polymer-supported, tetramethylammonium borohydrate, tetramethylammonium tetrahydroborate 1-, tetramethylammonium tetrahydroborate, tetra-methyl ammonium boron hydride, boron 1-; tetramethylazanium, n,n,n-trimethylmethanaminium tetrahydroborate 1-, ammonium, tetramethyl-, tetrahydroborate 1-, methanaminium, n,n,n-trimethyl-, tetrahydroborate 1- |
| PubChem CID | 6328186 |
| IUPAC Name | tetramethylazanium λ¹-boranuidylidene |
| SMILES | [B-].C[N+](C)(C)C |
Chemical Identifiers
| 16883-45-7 | |
| 84.96 | |
| TVQBZLPZXCVNEO-UHFFFAOYSA-N | |
| 6328186 | |
| [B-].C[N+](C)(C)C |
| C4H12BN | |
| MFCD00011778 | |
| tetramethylammonium borohydride, borohydride, polymer-supported, tetramethylammonium borohydrate, tetramethylammonium tetrahydroborate 1-, tetramethylammonium tetrahydroborate, tetra-methyl ammonium boron hydride, boron 1-; tetramethylazanium, n,n,n-trimethylmethanaminium tetrahydroborate 1-, ammonium, tetramethyl-, tetrahydroborate 1-, methanaminium, n,n,n-trimethyl-, tetrahydroborate 1- | |
| tetramethylazanium λ¹-boranuidylidene |
Specifications
| 16883-45-7 | |
| C4H12BN | |
| 5 g | |
| tetramethylammonium borohydride, borohydride, polymer-supported, tetramethylammonium borohydrate, tetramethylammonium tetrahydroborate 1-, tetramethylammonium tetrahydroborate, tetra-methyl ammonium boron hydride, boron 1-; tetramethylazanium, n,n,n-trimethylmethanaminium tetrahydroborate 1-, ammonium, tetramethyl-, tetrahydroborate 1-, methanaminium, n,n,n-trimethyl-, tetrahydroborate 1- | |
| [B-].C[N+](C)(C)C | |
| 84.96 | |
| 88.99 | |
| Crystalline Powder |
| White | |
| MFCD00011778 | |
| 3131 | |
| TVQBZLPZXCVNEO-UHFFFAOYSA-N | |
| tetramethylazanium λ¹-boranuidylidene | |
| 6328186 | |
| ≥90.0% (T) | |
| Tetramethylammonium Borohydride [Reducing Reagent] |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS8310000
TSCA : Yes