missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Borohydride 90.0+%, TCI America™

Catalog No. T08525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08525G 5 g
T085225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08525G Supplier TCI America Supplier No. T08525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16883-45-7
Molecular Formula C4H12BN
Molecular Weight (g/mol) 84.96
MDL Number MFCD00011778
InChI Key TVQBZLPZXCVNEO-UHFFFAOYSA-N
Synonym tetramethylammonium borohydride, borohydride, polymer-supported, tetramethylammonium borohydrate, tetramethylammonium tetrahydroborate 1-, tetramethylammonium tetrahydroborate, tetra-methyl ammonium boron hydride, boron 1-; tetramethylazanium, n,n,n-trimethylmethanaminium tetrahydroborate 1-, ammonium, tetramethyl-, tetrahydroborate 1-, methanaminium, n,n,n-trimethyl-, tetrahydroborate 1-
PubChem CID 6328186
IUPAC Name tetramethylazanium λ¹-boranuidylidene
SMILES [B-].C[N+](C)(C)C

Specifications

Color White
Quantity 5 g
UN Number 3131
Formula Weight 88.99
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Borohydride [Reducing Reagent]
EINECSNumber (2)-0186
RTECSNumber BS8310000
TSCA Yes