missing translation for 'onlineSavingsMsg'
Learn More

Tetramethoxysilane, 98%

Catalog No. p-7045184
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0971614 25 g
AAL0971636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0971614 Supplier Thermo Scientific Chemicals Supplier No. L0971614
Only null left

CAS: 681-84-5 | C4H12O4Si | 152.22 g/mol

Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

Solubility
Miscible with ethanol and organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.

Chemical Identifiers

CAS 681-84-5
Molecular Formula C4H12O4Si
Molecular Weight (g/mol) 152.22
MDL Number MFCD00008341
InChI Key LFQCEHFDDXELDD-UHFFFAOYSA-N
Synonym tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
PubChem CID 12682
IUPAC Name tetramethyl silicate
SMILES CO[Si](OC)(OC)OC

Specifications

Boiling Point 121°C to 122°C
Melting Point 4°C to 5°C
Quantity 25 g
UN Number UN2606
Beilstein 1699658
Flash Point 26°C (78°F)
Solubility Information Miscible with ethanol and organic solvents. Immiscible with water.
Formula Weight 152.22
Percent Purity 98%
Odor Ether-like
Refractive Index 1.368
Sensitivity Moisture sensitive
Density 1.032
Chemical Name or Material Tetramethoxysilane
Show More Show Less
Hazard Category H226-H315-H318-H330-H335
Hazard Statement GHS H Statement
H330-H318-H226-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ORTHOSILICATE
EINECSNumber 211-656-4
RTECSNumber VV9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only