missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethoxymethane 97.0+%, TCI America™
$614.13 - $2454.59
Chemical Identifiers
| CAS | 1850-14-2 |
|---|---|
| Molecular Formula | C5H12O4 |
| Molecular Weight (g/mol) | 136.147 |
| MDL Number | MFCD00008473 |
| InChI Key | AHJWSRRHTXRLAQ-UHFFFAOYSA-N |
| Synonym | tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc |
| PubChem CID | 74613 |
| IUPAC Name | tetramethoxymethane |
| SMILES | COC(OC)(OC)OC |
Chemical Identifiers
| 1850-14-2 | |
| 136.147 | |
| AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
| 74613 | |
| COC(OC)(OC)OC |
| C5H12O4 | |
| MFCD00008473 | |
| tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
| tetramethoxymethane |
Specifications
| 1850-14-2 | |
| 114°C | |
| MFCD00008473 | |
| 3272 | |
| AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
| tetramethoxymethane | |
| 74613 | |
| ≥97.0% (GC) | |
| Tetramethoxymethane |
| Colorless | |
| C5H12O4 | |
| 5 g | |
| tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
| COC(OC)(OC)OC | |
| 136.147 | |
| 136.15 | |
| Liquid |
Safety and Handling
TSCA : No