missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)platinum(0), 98%, Thermo Scientific™

Chemical Identifiers

CAS 14221-02-4
Molecular Formula C72H60P4Pt
Molecular Weight (g/mol) 1244.27
MDL Number MFCD00010014
InChI Key SYKXNRFLNZUGAJ-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
PubChem CID 11979705
IUPAC Name platinum;triphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223960010
SDS View Documents
Thermo Scientific Chemicals
223960010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC223960050
SDS View Documents
Thermo Scientific Chemicals
223960050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14221-02-4
1244.27
SYKXNRFLNZUGAJ-UHFFFAOYSA-N
11979705
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt]
C72H60P4Pt
MFCD00010014
tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
platinum;triphenylphosphane

Specifications

14221-02-4
Authentic
Glass bottle
[(C6H5)3P]4Pt
1g
tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
SYKXNRFLNZUGAJ-UHFFFAOYSA-N
platinum;triphenylphosphane
11979705
98%
97.5% min.
98%
C72H60P4Pt
MFCD00010014
15,300; 16,98; 04,33; 05,34; 09,45; 10,33; 14,36; 15,30; 16,29; 17,32; 06,570; 07,357; 09,450; 12,467; 13,294; 16,317
Solubility in water: insoluble.
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt]
1244.27
1244.27
Tetrakis(triphenylphosphine)platinum(0)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-087-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only