missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis), Thermo Scientific™

Chemical Identifiers

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Molecular Weight (g/mol) 1155.58
InChI Key NFHFRUOZVGFOOS-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
PubChem CID 11979704
IUPAC Name palladium;triphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441400010
SDS View Documents
Thermo Scientific Chemicals
441400010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441400050
SDS View Documents
Thermo Scientific Chemicals
441400050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
Specifications

Chemical Identifiers

14221-01-3
1155.58
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
palladium;triphenylphosphane
C72H60P4Pd
NFHFRUOZVGFOOS-UHFFFAOYSA-N
11979704
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]

Specifications

14221-01-3
Glass bottle
[(C6H5)3P]4Pd
15,9367
Solubility in water: insoluble.
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
1155.58
1155.58
Trace Metal Basis
99.9%
C72H60P4Pd
1g
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
NFHFRUOZVGFOOS-UHFFFAOYSA-N
palladium;triphenylphosphane
11979704
99.9%
Tetrakis(triphenylphosphine)palladium(0)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 238-086-9

SDS
Documents

Documents

RUO – Research Use Only