missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetrakis(Triphenylphosphine)Palladium(0)

Catalog No. 2166665G
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2166665G 5 g
21666625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 2166665G Supplier Sigma Aldrich Supplier No. 2166665G
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(triphenylphosphine)palladium(0) can be used as a catalyst in:Negishi coupling (eq. 1), Suzuki coupling (eq. 2), Stille coupling (eq. 3), Sonogashira coupling reaction (eq. 4), and Buchwald-Hartwig amination reaction (eq. 5)The carbonylation of vinyl iodides (eq. 6)The reduction reaction of aryl bromides (eq. 7)Carbon-tin bond formation (eq. 8)

Specifications

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Linear Formula Pd[(C6H5)3P]4
MDL Number MFCD00010012
Quantity 5 g
Synonym Palladium-tetrakis(triphenylphosphine); Pd(PPh3)4
Molecular Weight (g/mol) 1155.56 g/mol