missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(trimethylsilyl)silane 98.0+%, TCI America™
$55.92 - $171.49
Chemical Identifiers
| CAS | 4098-98-0 |
|---|---|
| Molecular Formula | C12H36Si5 |
| Molecular Weight (g/mol) | 320.85 |
| MDL Number | MFCD00054859 |
| InChI Key | BOJSDHZZKKYWAS-UHFFFAOYSA-N |
| Synonym | tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa |
| PubChem CID | 138115 |
| IUPAC Name | 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane |
| SMILES | C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
Chemical Identifiers
| 4098-98-0 | |
| 320.85 | |
| BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
| 138115 | |
| C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
| C12H36Si5 | |
| MFCD00054859 | |
| tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
| 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane |
Specifications
| 4098-98-0 | |
| C12H36Si5 | |
| 1 g | |
| BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane | |
| 138115 | |
| ≥98.0% (GC) | |
| Tetrakis(trimethylsilyl)silane |
| White-Yellow | |
| MFCD00054859 | |
| tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
| C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C | |
| 320.85 | |
| 320.85 | |
| Crystalline Powder |
Safety and Handling
TSCA : No