missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(trimethylsilyl)silane 98.0+%, TCI America™

Catalog No. T14645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14645G 5 g
T14641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14645G Supplier TCI America Supplier No. T14645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4098-98-0
Molecular Formula C12H36Si5
Molecular Weight (g/mol) 320.85
MDL Number MFCD00054859
InChI Key BOJSDHZZKKYWAS-UHFFFAOYSA-N
Synonym tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa
PubChem CID 138115
IUPAC Name 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 320.85
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(trimethylsilyl)silane