Learn More
Tetrakis(trimethylsilyl)silane, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 4098-98-0 |
|---|---|
| Molecular Formula | C12H36Si5 |
| Molecular Weight (g/mol) | 320.85 |
| MDL Number | MFCD00054859 |
| InChI Key | BOJSDHZZKKYWAS-UHFFFAOYSA-N |
| Synonym | tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa |
| PubChem CID | 138115 |
| IUPAC Name | tetrakis(trimethylsilyl)silane |
| SMILES | C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC293900010
|
Thermo Scientific Chemicals
293900010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC293900050
|
Thermo Scientific Chemicals
293900050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4098-98-0 | |
| 320.85 | |
| BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
| 138115 | |
| C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
| C12H36Si5 | |
| MFCD00054859 | |
| tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
| tetrakis(trimethylsilyl)silane |
Specifications
| 4098-98-0 | |
| 96% min. (GC) | |
| C12H36Si5 | |
| MFCD00054859 | |
| tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
| C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C | |
| 320.85 | |
| 320.84 | |
| Tetrakis(trimethylsilyl)silane |
| Authentic | |
| Glass Bottle | |
| [(CH3)3Si]4Si | |
| 1g | |
| BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
| tetrakis(trimethylsilyl)silane | |
| 138115 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air
GHS Signal Word: Warning
RUO â Research Use Only