missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(dimethylamino)titanium(IV) 97.0+%, TCI America™
$80.16 - $80.16
Chemical Identifiers
| CAS | 3275-24-9 |
|---|---|
| Molecular Formula | C8H24N4Ti |
| Molecular Weight (g/mol) | 224.18 |
| MDL Number | MFCD00014861 |
| InChI Key | MNWRORMXBIWXCI-UHFFFAOYSA-N |
| Synonym | tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva |
| PubChem CID | 123185 |
| IUPAC Name | titanium(4+) tetrakis(dimethylazanide) |
| SMILES | [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C |
Chemical Identifiers
| 3275-24-9 | |
| 224.18 | |
| MNWRORMXBIWXCI-UHFFFAOYSA-N | |
| 123185 | |
| [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C |
| C8H24N4Ti | |
| MFCD00014861 | |
| tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva | |
| titanium(4+) tetrakis(dimethylazanide) |
Specifications
| 3275-24-9 | |
| MFCD00014861 | |
| 3399 | |
| MNWRORMXBIWXCI-UHFFFAOYSA-N | |
| titanium(4+) tetrakis(dimethylazanide) | |
| 123185 | |
| ≥97.0% (T) | |
| Tetrakis(dimethylamino)titanium(IV) |
| C8H24N4Ti | |
| 1 g | |
| tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva | |
| [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C | |
| 224.18 | |
| 224.18 | |
| Liquid |
Safety and Handling
TSCA : Yes