missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis)

Catalog No. p-3523598
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC353560250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC353560250 Supplier Thermo Scientific Chemicals Supplier No. 353560250
Only null left

CAS: 3275-24-9 | C8H24N4Ti | 224.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 3275-24-9
Molecular Formula C8H24N4Ti
Molecular Weight (g/mol) 224.18
MDL Number MFCD00014861
InChI Key MNWRORMXBIWXCI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
PubChem CID 123185
SMILES [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C

Specifications

Color Yellow to Orange
Density 0.9470g/mL
Flash Point -30°C
Infrared Spectrum Authentic
Assay Percent Range 99.99%
Packaging Glass bottle
Linear Formula [(CH3)2N]4Ti
Quantity 25 g
Beilstein 04, IV, 128
Specific Gravity 0.947
IUPAC Name dimethylazanide;titanium(4+)
Formula Weight 224.21
Percent Purity 99.99%
Grade Trace Metal Basis
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)titanium
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose parti
EINECSNumber 221-904-3

RUO – Research Use Only