missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7058564
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4276009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4276009 Supplier Thermo Scientific Chemicals Supplier No. 04276009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Preparation of nitrides

Preparation of nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7344-40-3
Molecular Formula C8H24GeN4
Molecular Weight (g/mol) 248.938
MDL Number MFCD01863476
InChI Key JKUUTODNPMRHHZ-UHFFFAOYSA-N
Synonym germane, tetrakis dimethylamino, octamethylgermanetetramine, tetrakis dimethylamino germane, tetrakis dimethylamino germanium, dimethyl tris dimethylamino germyl amine
PubChem CID 16684231
IUPAC Name N-methyl-N-[tris(dimethylamino)germyl]methanamine
SMILES CN(C)[Ge](N(C)C)(N(C)C)N(C)C

Specifications

Melting Point 14°C
Color Colorless
Density 1.07 g/mL
Boiling Point 203°C
Flash Point 34°C (93°F)
Odor Amine-like
Linear Formula Ge[N(CH3)2]4
Quantity 10 g
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 248.9
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)germanium (IV)
Show More Show Less
Hazard Category H226-H261-H314-H335
Hazard Statement GHS H Statement
H261-H226-H314-H318
In contact with water releases flammable gas.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only