missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(dimethylamino)ethylene 95.0+%, TCI America™
$1,817.46 - $8,997.79
Chemical Identifiers
| CAS | 996-70-3 |
|---|---|
| Molecular Formula | C10H24N4 |
| Molecular Weight (g/mol) | 200.33 |
| MDL Number | MFCD00009601 |
| InChI Key | CBXRMKZFYQISIV-UHFFFAOYSA-N |
| Synonym | tetrakis dimethylamino ethylene, ethenetetramine, octamethyl, tdae, unii-p6737b68mn, n1,n1,n'1,n'1,n2,n2,n'2,n'2-octamethylethene-1,1,2,2-tetraamine, octamethylethylenetetramine, 1,1,2,2-ethenetetramine, n1,n1,n1',n1',n2,n2,n2',n2'-octamethyl, dimethyl 1,2,2-tris dimethylamino vinyl amine, acmc-209sdr, tetra dimethlamino ethylene |
| PubChem CID | 70455 |
| IUPAC Name | 1-N,1-N,1-N',1-N',2-N,2-N,2-N',2-N'-octamethylethene-1,1,2,2-tetramine |
| SMILES | CN(C)C(=C(N(C)C)N(C)C)N(C)C |
Chemical Identifiers
| 996-70-3 | |
| 200.33 | |
| CBXRMKZFYQISIV-UHFFFAOYSA-N | |
| 70455 | |
| CN(C)C(=C(N(C)C)N(C)C)N(C)C |
| C10H24N4 | |
| MFCD00009601 | |
| tetrakis dimethylamino ethylene, ethenetetramine, octamethyl, tdae, unii-p6737b68mn, n1,n1,n'1,n'1,n2,n2,n'2,n'2-octamethylethene-1,1,2,2-tetraamine, octamethylethylenetetramine, 1,1,2,2-ethenetetramine, n1,n1,n1',n1',n2,n2,n2',n2'-octamethyl, dimethyl 1,2,2-tris dimethylamino vinyl amine, acmc-209sdr, tetra dimethlamino ethylene | |
| 1-N,1-N,1-N',1-N',2-N,2-N,2-N',2-N'-octamethylethene-1,1,2,2-tetramine |
Specifications
| 996-70-3 | |
| 59°C | |
| MFCD00009601 | |
| 2734 | |
| CBXRMKZFYQISIV-UHFFFAOYSA-N | |
| 1-N,1-N,1-N',1-N',2-N,2-N,2-N',2-N'-octamethylethene-1,1,2,2-tetramine | |
| 70455 | |
| ≥95.0% (GC) | |
| Tetrakis(dimethylamino)ethylene |
| Yellow | |
| C10H24N4 | |
| 5 g | |
| tetrakis dimethylamino ethylene, ethenetetramine, octamethyl, tdae, unii-p6737b68mn, n1,n1,n'1,n'1,n2,n2,n'2,n'2-octamethylethene-1,1,2,2-tetraamine, octamethylethylenetetramine, 1,1,2,2-ethenetetramine, n1,n1,n1',n1',n2,n2,n2',n2'-octamethyl, dimethyl 1,2,2-tris dimethylamino vinyl amine, acmc-209sdr, tetra dimethlamino ethylene | |
| CN(C)C(=C(N(C)C)N(C)C)N(C)C | |
| 200.33 | |
| 200.33 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator