missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(dimethylamino)ethylene 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 59°C |
| Quantity | 5 g |
| UN Number | 2734 |
| Formula Weight | 200.33 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tetrakis(dimethylamino)ethylene |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |