missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate 95.0+%, TCI America™
$218.10 - $1291.79
Chemical Identifiers
| CAS | 126150-97-8 |
|---|---|
| Molecular Formula | C34H40N2O18 |
| Molecular Weight (g/mol) | 764.69 |
| MDL Number | MFCD00036696 |
| InChI Key | YJIYWYAMZFVECX-UHFFFAOYSA-N |
| Synonym | bapta-am, bapta/am, bapta am, tetrakis acetoxymethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrakis acetoxymethyl ester, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 2-2-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy phenyl amino acetate, 1,2-bis o-aminophenoxy ethane-n,n,n′,n′-tetraacetic acid tetra acetoxymethyl ester, acmc-20d2ub, bapta, am, bapta-am hplc |
| PubChem CID | 2293 |
| IUPAC Name | acetyloxymethyl 2-[N-[2-(acetyloxymethoxy)-2-oxoethyl]-2-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]anilino]acetate |
| SMILES | CC(=O)OCOC(=O)CN(CC(=O)OCOC(=O)C)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T284520MG
|
TCI America
T284520MG |
20 mg |
Each for $218.10
|
|
|||||
|
T2845200MG
|
TCI America
T2845200MG |
200 mg |
Each for $1,291.79
|
|
|||||
Chemical Identifiers
| 126150-97-8 | |
| 764.69 | |
| YJIYWYAMZFVECX-UHFFFAOYSA-N | |
| 2293 | |
| CC(=O)OCOC(=O)CN(CC(=O)OCOC(=O)C)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C |
| C34H40N2O18 | |
| MFCD00036696 | |
| bapta-am, bapta/am, bapta am, tetrakis acetoxymethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrakis acetoxymethyl ester, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 2-2-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy phenyl amino acetate, 1,2-bis o-aminophenoxy ethane-n,n,n′,n′-tetraacetic acid tetra acetoxymethyl ester, acmc-20d2ub, bapta, am, bapta-am hplc | |
| acetyloxymethyl 2-[N-[2-(acetyloxymethoxy)-2-oxoethyl]-2-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]anilino]acetate |
Specifications
| 126150-97-8 | |
| White | |
| MFCD00036696 | |
| bapta-am, bapta/am, bapta am, tetrakis acetoxymethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrakis acetoxymethyl ester, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 2-2-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy phenyl amino acetate, 1,2-bis o-aminophenoxy ethane-n,n,n′,n′-tetraacetic acid tetra acetoxymethyl ester, acmc-20d2ub, bapta, am, bapta-am hplc | |
| CC(=O)OCOC(=O)CN(CC(=O)OCOC(=O)C)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C | |
| 764.69 | |
| 764.69 | |
| Crystalline Powder |
| 98°C | |
| C34H40N2O18 | |
| 20 mg | |
| YJIYWYAMZFVECX-UHFFFAOYSA-N | |
| acetyloxymethyl 2-[N-[2-(acetyloxymethoxy)-2-oxoethyl]-2-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]anilino]acetate | |
| 2293 | |
| ≥95.0% (HPLC) | |
| Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer