missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate, Pd 23.5%

Catalog No. AA3944601
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3944601 0.25 g
AA3944603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3944601 Supplier Thermo Scientific Chemicals Supplier No. 03944601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21797-13-7 | C8H12B2F8N4Pd | 444.24 g/mol

Acts as a precursor for:

  • Synthesis of dendritic SCS-pincer palladium complexes.
  • Palladium complexes of click ligands.
  • Dipalladium catalysts for use in Heck cross-coupling5, Suzuki cross-coupling, and aldehyde olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21797-13-7
Molecular Formula C8H12B2F8N4Pd
Molecular Weight (g/mol) 444.24
MDL Number MFCD00043297
InChI Key YWMRPVUMBTVUEX-UHFFFAOYSA-N
Synonym tetrakis acetonitrile palladium ii tetrafluoroborate, acetonitrile; palladium 2+ ; ditetrafluoroborate, pd mecn 4 bf4 2, tetrakis acetonitrile palladium ii, palladium ii tetrafluoroborate tetraacetonitrile complex
PubChem CID 2734560
SMILES [Pd++].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Melting Point 230°C (decomposition)
Quantity 0.25 g
UN Number UN2811
Sensitivity Air Sensitive
Solubility Information Soluble in water
IUPAC Name palladium(2+) tetrakis(acetonitrile) bis(tetrafluoroboranuide)
Formula Weight 444.22
Concentration or Composition (by Analyte or Components) Pd 23.5%
Physical Form Powder
Chemical Name or Material Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Hazard Category H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P271-P304+P340-P312
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only