missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(acetonitrile)palladium(II) Bis(trifluoromethanesulfonate) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 568.76 |
| Percent Purity | ≥95.0% (N,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetrakis(acetonitrile)palladium(II) Bis(trifluoromethanesulfonate) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |