missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(4-ethynylphenyl)methane 98.0+%, TCI America™

Catalog No. T3151100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3151100MG 100 mg
T31511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3151100MG Supplier TCI America Supplier No. T3151100MG

Chemical Identifiers

CAS 177991-01-4
Molecular Formula C33H20
Molecular Weight (g/mol) 416.52
MDL Number MFCD24873189
InChI Key JGUCYLISNORQFW-UHFFFAOYSA-N
PubChem CID 12157742
IUPAC Name 1-ethynyl-4-[tris(4-ethynylphenyl)methyl]benzene
SMILES C#CC1=CC=C(C=C1)C(C1=CC=C(C=C1)C#C)(C1=CC=C(C=C1)C#C)C1=CC=C(C=C1)C#C

Specifications

Melting Point 300°C
Color Yellow
Quantity 100 mg
Formula Weight 416.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(4-ethynylphenyl)methane