missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051819
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4202806 5 g
AA4202814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4202806 Supplier Thermo Scientific Chemicals Supplier No. 04202806
Only null left

MOCVD of zirconium

MOCVD of zirconium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18865-74-2
Molecular Formula C44H80O8Zr
Molecular Weight (g/mol) 828.34
MDL Number MFCD00145380,MFCD00145380
InChI Key ANBXKEDJQLYCLX-UHFFFAOYSA-N
Synonym tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
PubChem CID 50919870
IUPAC Name 5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
SMILES [Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C

Specifications

Melting Point 318°C to 320°C
Boiling Point 180°C (0.1 mmHg)(sublimation)
Quantity 5 g
Formula Weight 824.3
Percent Purity 99.99%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only