missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals
$1253.37 - $1253.37
Chemical Identifiers
| CAS | 63370-90-1 |
|---|---|
| Molecular Formula | C44H76HfO8 |
| Molecular Weight (g/mol) | 911.574 |
| MDL Number | MFCD01863471 |
| InChI Key | GPCQYWUYUFMJFR-UHFFFAOYSA-N |
| Synonym | hafnium tetramethylheptanedionate, tetrakis dipivaloylmethanato hafnium |
| PubChem CID | 121233202 |
| SMILES | CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.[Hf+4] |
Description
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV) is used as an intermediate to fabricate capacitor devices on the nano scale.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 63370-90-1 | |
| 911.574 | |
| GPCQYWUYUFMJFR-UHFFFAOYSA-N | |
| 121233202 |
| C44H76HfO8 | |
| MFCD01863471 | |
| hafnium tetramethylheptanedionate, tetrakis dipivaloylmethanato hafnium | |
| CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.[Hf+4] |
Specifications
| 63370-90-1 | |
| C44H76HfO8 | |
| 5 g | |
| Soluble in hexane,toluene,hydrocarbons,alcohols,ketones and esters. Insoluble in water. | |
| CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.[Hf+4] | |
| 121233202 | |
| 99.7% | |
| Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV) |
| 315°C | |
| MFCD01863471 | |
| hafnium tetramethylheptanedionate, tetrakis dipivaloylmethanato hafnium | |
| GPCQYWUYUFMJFR-UHFFFAOYSA-N | |
| 911.574 | |
| 910.5 | |
| Powder | |
| (metals basis) |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only