missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(1-pyrazolyl)borate, potassium salt, 95%

Catalog No. AC300160050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300160050 5 g Glass bottle
AC300160010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC300160050 Supplier Thermo Scientific Chemicals Supplier No. 300160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14782-58-2 | C12H12BKN8 | 318.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14782-58-2
Molecular Formula C12H12BKN8
Molecular Weight (g/mol) 318.2
MDL Number MFCD00800557
InChI Key RKBGLZQVHBLUBM-UHFFFAOYSA-N
Synonym tetrakis 1-pyrazolyl borate, potassium tetrakis pyrazol-1-yloxy boranuide
PubChem CID 92043288
IUPAC Name potassium;tetra(pyrazol-1-yloxy)boranuide
SMILES [B-](ON1C=CC=N1)(ON2C=CC=N2)(ON3C=CC=N3)ON4C=CC=N4.[K+]

Specifications

Melting Point 253°C to 254°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohols,dmf,thf
Formula Weight 318.2
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Show More Show Less

RUO – Research Use Only