missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl Orthotitanate, TCI America™

Catalog No. T013325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013325G 25 g
T0133500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T013325G Supplier TCI America Supplier No. T013325G

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.219
MDL Number MFCD00008871
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
UN Number 2413
Formula Weight 284.22
Physical Form Liquid
Chemical Name or Material Tetraisopropyl Orthotitanate
EINECSNumber (2)-0228&(2)-2150
RTECSNumber NT8060000
TSCA Yes