missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl Methylenediphosphonate 97.0+%, TCI America™

Catalog No. M131925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M131925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M131925G Supplier TCI America Supplier No. M131925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1660-95-3
Molecular Formula C13H30O6P2
Molecular Weight (g/mol) 344.325
MDL Number MFCD00015021
InChI Key ODTQUKVFOLFLIQ-UHFFFAOYSA-N
Synonym tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847
PubChem CID 74266
IUPAC Name 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane
SMILES CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C

Specifications

Color Colorless
Quantity 25 g
Formula Weight 344.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraisopropyl Methylenediphosphonate