missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl methylenediphosphonate, 98%

Catalog No. AAA1348430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348430 250 g
AAA1348409 10 g
AAA1348418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1348430 Supplier Thermo Scientific Chemicals Supplier No. A1348430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1660-95-3 | C13H30O6P2 | 344.325 g/mol

Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 1660-95-3
Molecular Formula C13H30O6P2
Molecular Weight (g/mol) 344.325
MDL Number MFCD00015021
InChI Key ODTQUKVFOLFLIQ-UHFFFAOYSA-N
Synonym tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847
PubChem CID 74266
IUPAC Name 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane
SMILES CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C

Specifications

Density 1.08
Boiling Point 122°C to 124°C (1.5 mmHg)
Flash Point 160°C (320°F)
Refractive Index 1.434
Quantity 250 g
Beilstein 1080180
Solubility Information Insoluble in water.
Formula Weight 344.33
Percent Purity 98%
Chemical Name or Material Tetraisopropyl methylenediphosphonate
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 216-765-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only