missing translation for 'onlineSavingsMsg'
Learn More

Tetraiodo-2-sulfobenzoic Anhydride 95.0+%, TCI America™

Catalog No. T013125G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013125G 25 g
T01311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T013125G Supplier TCI America Supplier No. T013125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1745-83-1
Molecular Formula C7I4O4S
Molecular Weight (g/mol) 687.751
MDL Number MFCD00059713
InChI Key UQDQTEHMZFWZSX-UHFFFAOYSA-N
PubChem CID 636253
IUPAC Name 4,5,6,7-tetraiodo-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C12=C(C(=C(C(=C1I)I)I)I)S(=O)(=O)OC2=O

Specifications

Color Yellow
Quantity 25 g
UN Number 1759
Formula Weight 687.75
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraiodo-2-sulfobenzoic Anhydride