missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydroxydiboron, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Assay Percent Range | 95% |
| Linear Formula | (HO)2BB(OH)2 |
| Quantity | 5 g |
| Solubility Information | Solubility aq. soln.: .. Other solubilities: soluble in methanol |
| Formula Weight | 89.65 |
| Chemical Name or Material | Tetrahydroxydiboron |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention. |
RUO – Research Use Only