missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrothiopyran, 98%
Description
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
Specifications
Specifications
| Density | 0.988 |
| Boiling Point | 140°C to 142°C |
| Flash Point | 21°C (69°F) |
| Odor | Unpleasant |
| Refractive Index | 1.506 |
| Quantity | 1 g |
| UN Number | UN1993 |
| Beilstein | 102437 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 102.2 |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 216-561-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only