missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran, 98%

Catalog No. AAL0306303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0306303 1 g
AAL0306306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0306303 Supplier Thermo Scientific Chemicals Supplier No. L0306303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1613-51-0 | C5H10S | 102.195 g/mol

It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.

Chemical Identifiers

CAS 1613-51-0
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.195
MDL Number MFCD00006662
InChI Key YPWFISCTZQNZAU-UHFFFAOYSA-N
Synonym pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
PubChem CID 15367
IUPAC Name thiane
SMILES C1CCSCC1

Specifications

Density 0.988
Boiling Point 140°C to 142°C
Flash Point 21°C (69°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 1 g
UN Number UN1993
Beilstein 102437
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 102.2
Percent Purity 98%
Chemical Name or Material Tetrahydrothiopyran
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-561-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only