missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran 98.0+%, TCI America™

Catalog No. T0110500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0110500ML 500 mL
T011025ML 25 mL
T0110100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0110500ML Supplier TCI America Supplier No. T0110500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-68-7
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00006585
InChI Key DHXVGJBLRPWPCS-UHFFFAOYSA-N
Synonym tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pye
PubChem CID 8894
ChEBI CHEBI:46941
IUPAC Name oxane
SMILES C1CCOCC1

Specifications

Color Yellow
Boiling Point 88°C
Quantity 500 mL
UN Number 1993
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydropyran
EINECSNumber (5)-5761
TSCA Yes