missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran, 98+%

CAS: 142-68-7 | C5H10O | 86.134 g/mol

$100.99 - $100.99

Chemical Identifiers

CAS 142-68-7
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00006585
InChI Key DHXVGJBLRPWPCS-UHFFFAOYSA-N
Synonym tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pye
PubChem CID 8894
ChEBI CHEBI:46941
IUPAC Name oxane
SMILES C1CCOCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA13392AE
View Documents
Thermo Scientific Chemicals
A13392AE
100 mL
Each for $100.99
Only null left
 
Description

Description

It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Oxidizing agents, Strong acids. May form peroxides in the absence of inhibitors. Light sensitive
Specifications

Chemical Identifiers

142-68-7
86.134
DHXVGJBLRPWPCS-UHFFFAOYSA-N
8894
oxane
C5H10O
MFCD00006585
tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pye
CHEBI:46941
C1CCOCC1

Specifications

142-68-7
0.882
−15°C (5°F)
1.421
100 mL
102436
tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pye
DHXVGJBLRPWPCS-UHFFFAOYSA-N
oxane
8894
86.14
Tetrahydropyran
-49°C
87°C to 89°C
C5H10O
MFCD00006585
UN3271
14,9217
It is soluble in water.
C1CCOCC1
86.134
CHEBI:46941
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 205-552-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only