missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-sulfonyl chloride, 97%

Catalog No. p-4228912
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429440010 1 g Glass bottle
AC429440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429440010 Supplier Thermo Scientific Chemicals Supplier No. 429440010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 338453-21-7 | C5H9ClO3S | 184.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 338453-21-7
Molecular Formula C5H9ClO3S
Molecular Weight (g/mol) 184.64
InChI Key QQSBPTQNEOJFBO-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-sulfonyl chloride, tetrahydropyran-4-sulfonyl chloride, tetrahydro-pyran-4-sulfonyl chloride, 2h-pyran-4-sulfonylchloride, tetrahydro, tetrahydropyran-4-sulfonylchloride, tetrahydropyran-4-sulphonyl chloride, 2h-pyran-4-sulfonylchloride,tetrahydro, n-5-bromo-2-methoxybenzyl cyclohexanamine hydrobromide
PubChem CID 22637030
IUPAC Name oxane-4-sulfonyl chloride
SMILES C1COCCC1S(=O)(=O)Cl

Specifications

Melting Point 36.0°C to 40.0°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 184.64
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Tetrahydropyran-4-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Store in a dry place.
Store in a closed container.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with

RUO – Research Use Only