missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydropyran-4-methanol 97.0+%, TCI America™
$234.87 - $234.87
Chemical Identifiers
| CAS | 14774-37-9 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00457804 |
| InChI Key | YSNVSVCWTBLLRW-UHFFFAOYSA-N |
| Synonym | tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol |
| PubChem CID | 2773573 |
| IUPAC Name | (oxan-4-yl)methanol |
| SMILES | OCC1CCOCC1 |
Chemical Identifiers
| 14774-37-9 | |
| 116.16 | |
| YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
| 2773573 | |
| OCC1CCOCC1 |
| C6H12O2 | |
| MFCD00457804 | |
| tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
| (oxan-4-yl)methanol |
Specifications
| 14774-37-9 | |
| 110°C | |
| MFCD00457804 | |
| tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
| OCC1CCOCC1 | |
| 116.16 | |
| 116.16 | |
| Liquid |
| Yellow | |
| C6H12O2 | |
| 1 g | |
| YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
| (oxan-4-yl)methanol | |
| 2773573 | |
| ≥97.0% (GC) | |
| Tetrahydropyran-4-methanol |
Safety and Handling
TSCA : No