missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. T24935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24935G 5 g
T249325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24935G Supplier TCI America Supplier No. T24935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00031016
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha
PubChem CID 219302
IUPAC Name oxane-4-carboxylic acid
SMILES C1COCCC1C(=O)O

Specifications

Melting Point 89°C
Color White
Boiling Point 115°C
Quantity 5 g
Formula Weight 130.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydropyran-4-carboxylic Acid