missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-carboxylic acid, 98%

Catalog No. p-7055696
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6434006 5 g
AAH6434014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6434006 Supplier Thermo Scientific Chemicals Supplier No. H6434006

CAS: 5337-03-1 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00031016
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha
PubChem CID 219302
IUPAC Name oxane-4-carboxylic acid
SMILES C1COCCC1C(=O)O

Specifications

Melting Point 87°C to 89°C
Color White
Quantity 5 g
Beilstein 112249
Formula Weight 130.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Tetrahydropyran-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only