Learn More
Tetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 456720050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Tetrahydropyran-4-carboxylic acid | |
| 115.0°C (20.0 mmHg) | |
| C6H10O3 | |
| tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha | |
| AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
| oxane-4-carboxylic acid | |
| 219302 | |
| 97% |
| 5337-03-1 | |
| Authentic | |
| 5g | |
| Solubility in water: .. Other solubilities: soluble in methanol | |
| C1COCCC1C(=O)O | |
| 130.14 | |
| 130.14 |
Chemical Identifiers
| 5337-03-1 | |
| 130.14 | |
| tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha | |
| oxane-4-carboxylic acid |
| C6H10O3 | |
| AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
| 219302 | |
| C1COCCC1C(=O)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only