missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-carbothioamide, 90%, Thermo Scientific™

Chemical Identifiers

CAS 88571-77-1
Molecular Formula C6H11NOS
Molecular Weight (g/mol) 145.22
MDL Number MFCD10700048
InChI Key ZUONFEFOQCUTDW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carbothioamide, tetrahydropyran-4-carbothioamide, 2h-pyran-4-carbothioamide,tetrahydro, 2h-pyran-4-carbothioamide, tetrahydro, acmc-20fauh, oxane-4-carboimidothioic acid, 4-carbamothioyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-carbothioic acid amide, 2h-3,4,5,6-tetrahydropyran-4-ylaminomethane-1-thione
PubChem CID 13197204
IUPAC Name oxane-4-carbothioamide
SMILES C1COCCC1C(=S)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC29918CB
SDS View Documents
Thermo Scientific Chemicals
CC29918CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC29918DA
SDS View Documents
Thermo Scientific Chemicals
CC29918DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

88571-77-1
145.22
ZUONFEFOQCUTDW-UHFFFAOYSA-N
13197204
C1COCCC1C(=S)N
C6H11NOS
MFCD10700048
tetrahydro-2h-pyran-4-carbothioamide, tetrahydropyran-4-carbothioamide, 2h-pyran-4-carbothioamide,tetrahydro, 2h-pyran-4-carbothioamide, tetrahydro, acmc-20fauh, oxane-4-carboimidothioic acid, 4-carbamothioyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-carbothioic acid amide, 2h-3,4,5,6-tetrahydropyran-4-ylaminomethane-1-thione
oxane-4-carbothioamide

Specifications

88571-77-1
100.0
Amber Glass Bottle
MFCD10700048
tetrahydro-2h-pyran-4-carbothioamide, tetrahydropyran-4-carbothioamide, 2h-pyran-4-carbothioamide,tetrahydro, 2h-pyran-4-carbothioamide, tetrahydro, acmc-20fauh, oxane-4-carboimidothioic acid, 4-carbamothioyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-carbothioic acid amide, 2h-3,4,5,6-tetrahydropyran-4-ylaminomethane-1-thione
C1COCCC1C(=S)N
145.22
145.22
Tetrahydropyran-4-carbothioamide
96.5
97%
C6H11NOS
250mg
ZUONFEFOQCUTDW-UHFFFAOYSA-N
oxane-4-carbothioamide
13197204
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'