missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-ylmethylamine, 97%, Thermo Scientific™

$260.60 - $260.60

Chemical Identifiers

CAS 683233-12-7
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD06738971
InChI Key NGHRATGQJXKQIQ-UHFFFAOYSA-N
Synonym oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride
PubChem CID 43811037
IUPAC Name oxan-2-ylmethanamine;hydrochloride
SMILES C1CCOC(C1)CN.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC41213CB
SDS View Documents
Thermo Scientific Chemicals
CC41213CB
250 mg Amber Glass Bottle
Each for $260.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

683233-12-7
151.634
NGHRATGQJXKQIQ-UHFFFAOYSA-N
43811037
C1CCOC(C1)CN.Cl
C6H14ClNO
MFCD06738971
oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride
oxan-2-ylmethanamine;hydrochloride

Specifications

683233-12-7
100.0
Amber Glass Bottle
MFCD06738971
oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride
C1CCOC(C1)CN.Cl
151.634
151.64
Tetrahydropyran-2-ylmethylamine hydrochloride
94.5
95%
C6H14ClNO
250 mg
NGHRATGQJXKQIQ-UHFFFAOYSA-N
oxan-2-ylmethanamine;hydrochloride
43811037
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'