missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-ylmethylamine, 97%, Thermo Scientific™

Catalog No. CC41213CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC41213CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC41213CB Supplier Thermo Scientific Chemicals Supplier No. CC41213CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 683233-12-7
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD06738971
InChI Key NGHRATGQJXKQIQ-UHFFFAOYSA-N
Synonym oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride
PubChem CID 43811037
IUPAC Name oxan-2-ylmethanamine;hydrochloride
SMILES C1CCOC(C1)CN.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 151.64
Percent Purity 95%
Chemical Name or Material Tetrahydropyran-2-ylmethylamine hydrochloride
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.