missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-methanol 96.0+%, TCI America™

Catalog No. T09525ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09525ML 5 mL
T095225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09525ML Supplier TCI America Supplier No. T09525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-72-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00006624
InChI Key ROTONRWJLXYJBD-UHFFFAOYSA-N
Synonym tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
PubChem CID 7524
IUPAC Name oxan-2-ylmethanol
SMILES C1CCOC(C1)CO

Specifications

Color Colorless
Boiling Point 187°C
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydropyran-2-methanol
RTECSNumber UQ0175000
TSCA No