Learn More
Tetrahydropyran-2-carboxylic acid, 97%, Thermo Scientific™
$227.23 - $227.23
Chemical Identifiers
| CAS | 51673-83-7 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD07779239 |
| InChI Key | MQAYFGXOFCEZRW-UHFFFAOYSA-N |
| Synonym | tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid |
| PubChem CID | 10964532 |
| IUPAC Name | oxane-2-carboxylic acid |
| SMILES | C1CCOC(C1)C(=O)O |
Chemical Identifiers
| 51673-83-7 | |
| 130.143 | |
| MQAYFGXOFCEZRW-UHFFFAOYSA-N | |
| 10964532 | |
| C1CCOC(C1)C(=O)O |
| C6H10O3 | |
| MFCD07779239 | |
| tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid | |
| oxane-2-carboxylic acid |
Specifications
| 51673-83-7 | |
| 100.0 | |
| C6H10O3 | |
| 250 mg | |
| MQAYFGXOFCEZRW-UHFFFAOYSA-N | |
| oxane-2-carboxylic acid | |
| 10964532 | |
| 95% |
| 96.5 | |
| 95% | |
| MFCD07779239 | |
| tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid | |
| C1CCOC(C1)C(=O)O | |
| 130.143 | |
| 130.14 | |
| Tetrahydropyran-2-carboxylic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.