missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrolinalool 98.0+%, TCI America™

Catalog No. T099125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T099125ML 25 mL
T0991500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T099125ML Supplier TCI America Supplier No. T099125ML
Only null left

Chemical Identifiers

CAS 78-69-3
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00004482
InChI Key DLHQZZUEERVIGQ-UHFFFAOYSA-N
Synonym tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670
PubChem CID 6548
ChEBI CHEBI:84242
IUPAC Name 3,7-dimethyloctan-3-ol
SMILES CCC(C)(CCCC(C)C)O

Specifications

Color Colorless
Boiling Point 196°C
Quantity 25 mL
Formula Weight 158.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydrolinalool
EINECSNumber (2)-0217
RTECSNumber RH0905000
TSCA Yes