missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfurylamine 98.0+%, TCI America™

Catalog No. T107225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T107225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T107225ML Supplier TCI America Supplier No. T107225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4795-29-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005373
InChI Key YNOGYQAEJGADFJ-UHFFFAOYNA-N
Synonym tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine
PubChem CID 253298
IUPAC Name 1-(oxolan-2-yl)methanamine
SMILES NCC1CCCO1

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2943
Boiling Point 155°C
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Tetrahydrofurfurylamine
EINECSNumber (9)-0854
RTECSNumber LV0175000
TSCA Yes