missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfurylamine, 97%

Catalog No. AC157825000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157825000 500 g Glass bottle
AC157820050 5 g Glass bottle
AC157821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157825000 Supplier Thermo Scientific Chemicals Supplier No. 157825000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4795-29-3 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4795-29-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005373
InChI Key YNOGYQAEJGADFJ-UHFFFAOYNA-N
Synonym tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine
PubChem CID 253298
IUPAC Name oxolan-2-ylmethanamine
SMILES NCC1CCCO1

Specifications

Color Colorless to Yellow
Density 0.9800g/mL
Quantity 500 g
Boiling Point 153.0°C to 154.0°C (744.0mmHg), 153.0°C to 154.0°C (744.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4540 to 1.4560
Specific Gravity 0.98
Assay Percent Range 97%
Viscosity 5 mPa.s (20°C)
Beilstein 18, II, 415
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in alcohol and ether
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material Tetrahydrofurfurylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-351-9
RTECSNumber LV0175000
TSCA TSCA

RUO – Research Use Only