missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Propionate 97.0+%, TCI America™
$714.68 - $714.68
Chemical Identifiers
| CAS | 637-65-0 |
|---|---|
| Molecular Formula | C8H14O3 |
| Molecular Weight (g/mol) | 158.197 |
| MDL Number | MFCD00047080 |
| InChI Key | FMKCDSXOYLTWBR-UHFFFAOYSA-N |
| Synonym | Propionic Acid Tetrahydrofurfuryl Ester |
| PubChem CID | 61183 |
| IUPAC Name | oxolan-2-ylmethyl propanoate |
| SMILES | CCC(=O)OCC1CCCO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T1016500ML
|
TCI America
T1016500ML |
500 mL |
Each for $714.68
|
|
|||||
Chemical Identifiers
| 637-65-0 | |
| 158.197 | |
| FMKCDSXOYLTWBR-UHFFFAOYSA-N | |
| 61183 | |
| CCC(=O)OCC1CCCO1 |
| C8H14O3 | |
| MFCD00047080 | |
| Propionic Acid Tetrahydrofurfuryl Ester | |
| oxolan-2-ylmethyl propanoate |
Specifications
| 637-65-0 | |
| 205°C | |
| Clear Liquid at 20°C | |
| MFCD00047080 | |
| FMKCDSXOYLTWBR-UHFFFAOYSA-N | |
| oxolan-2-ylmethyl propanoate | |
| 61183 | |
| ≥97.0% (GC) |
| Colorless | |
| 500 mL | |
| C8H14O3 | |
| Propionic Acid Tetrahydrofurfuryl Ester | |
| CCC(=O)OCC1CCCO1 | |
| 158.197 | |
| 158.20 | |
| Tetrahydrofurfuryl Propionate |
Safety and Handling
TSCA : Yes