missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Propionate 97.0+%, TCI America™

Catalog No. T1016500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1016500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1016500ML Supplier TCI America Supplier No. T1016500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-65-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00047080
InChI Key FMKCDSXOYLTWBR-UHFFFAOYSA-N
Synonym Propionic Acid Tetrahydrofurfuryl Ester
PubChem CID 61183
IUPAC Name oxolan-2-ylmethyl propanoate
SMILES CCC(=O)OCC1CCCO1

Specifications

Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
Boiling Point 205°C
Formula Weight 158.20
Percent Purity ≥97.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Propionate